Workrite FR FR/CP™ Lab Coat, men's
Provides dual hazard protection with flame resistance and chemical protection. Workrite FR FR/CP™ Lab Coats are lightweight, breathable, and comfortable to wear. They employ Nomex™ IIIA fabric to prevent ignition and ShieldTEC™ to repel small amounts of liquid chemicals.
Manufacturer: Workrite FR 353CH45RB4L 0L
 View more versions of this product
Catalog No. 19-800-062
  Check Availability
Add to Cart
    Specifications
    Finish Westex ShieldCXP Chemical Splash Protection
    Material Nomex IIIA fabric
    No. of Pockets 3
    Size 4XL
    Gender Men's
    Style Long
    Weight (English) 4.5 oz.
    Weight (Metric) 128g
    Closure Type Snap
    Color Royal blue
    Cuff Style Knit (FR but not CP)
    • Nomex IIIA fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame
    • ShieldCXP repels small amounts of liquid chemicals that are either dropped or splashed on the fabric
    • Nomex knit cuffs are fire resistant but do not provide chemical protection - these are typically covered by gloves
    • Black lay-down collar
    • Concealed snap front closure
    • Splash-resistant side pass-through for easy inner pocket access
    • Pockets include one double-stitched chest pocket and two large double-stitched lower front pockets with snap flaps
    • Available in a variety of sizes with regular and long variants
    Certifications
    NFPA 2112 UL-Certified
  • Description & Specifications
Related Products
Product Content Feedback