Learn More
Workrite FR FR/CP™ Lab Coat, men's
Provides dual hazard protection with flame resistance and chemical protection. Workrite FR FR/CP™ Lab Coats are lightweight, breathable, and comfortable to wear. They employ Nomex™ IIIA fabric to prevent ignition and ShieldCXP™ to repel small amounts of liquid chemicals.
Manufacturer:  Workrite FR 353CH45RB5L 0L
 View more versions of this product
Catalog No. 19-800-064
  Check Availability
Add to cart
Specifications
Description
  • Nomex IIIA fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame
  • ShieldCXP repels small amounts of liquid chemicals that are either dropped or splashed on the fabric
  • Nomex knit cuffs are fire resistant but do not provide chemical protection - these are typically covered by gloves
  • Black lay-down collar
  • Concealed snap front closure
  • Splash-resistant side pass-through for easy inner pocket access
  • Pockets include one double-stitched chest pocket and two large double-stitched lower front pockets with snap flaps
  • Available in a variety of sizes with regular and long variants
Certifications
NFPA 2112 UL-Certified
Description & Specifications
Specifications
Snap
Knit (FR but not CP)
3
Westex ShieldCXP Chemical Splash Protection
Long
128g
Royal blue
Lightweight Nomex IIIA Fabric
5X-Large
Men's
4.5 oz.