Workrite FR FR/CP™ Lab Coat, women's

Provide dual hazard protection with flame resistance and chemical protection. Workrite FR FR/CP™ Lab Coats are lightweight, breathable, and comfortable to wear. They employ Nomex™ IIIA fabric to prevent ignition and ShieldTEC™ to repel small amounts of liquid chemicals.

Manufacturer: Workrite FR 354CH45RBXL 00

 View more versions of this product

Catalog No. 19-800-069

  Check Availability
Add to Cart

    Specifications

    Finish Westex ShieldCXP Chemical Splash Protection
    No. of Pockets 3
    Length (English) Sleeve 34 in.
    Size (English) Chest 42.5 in.
    Size (Metric) Chest 108cm
    Weight (Metric) 128g
    Cuff Style Knit (FR but not CP)
    Material Nomex IIIA fabric
    Size XL
    Gender Women's
    Length (Metric) Sleeve 86.4cm
    Style Regular
    Weight (English) 4.5 oz.
    Closure Type Snap
    Color Royal blue

    • Nomex IIIA fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame
    • ShieldCXP repels small amounts of liquid chemicals that are either dropped or splashed on the fabric
    • Nomex knit cuffs are fire resistant but do not provide chemical protection - these are typically covered by gloves
    • Black lay-down collar
    • Concealed snap front closure
    • Splash-resistant side pass-through for easy inner pocket access
    • Pockets include one double-stitched chest pocket and two large double-stitched lower front pockets with snap flaps
    • Available in a variety of sizes
    • Includes a waistband in the back for a more tailored fit

    Certifications

    NFPA 2112 UL-Certified

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit