Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRA-1/FOSL1 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI12F9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271351JF549

Catalog No. N271351J549


Add to cart

Description

Description

FRA-1/FOSL1 Monoclonal antibody specifically detects FRA-1/FOSL1 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

FRA-1/FOSL1
OTI12F9
Immunohistochemistry
Affinity Purified, Conjugated
FOSL1
Full-length protein expressed in 293T cell transfected with human FRA-1/FOSL1 expression vector.
Immunogen affinity purified
Cell Cycle and Replication, Wnt Signaling Pathway
Primary
8061.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
FOS-like antigen 1, FOS-like antigen-1, fos-related antigen 1, FRA, FRA1, FRA-1
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit