Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRA2 Mouse anti-Human, Clone: PCRP-FOSL2-1B1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30759110UG

 View more versions of this product

Catalog No. NB122655


Add to cart

Description

Description

FRA2 Monoclonal antibody specifically detects FRA2 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein Array
Specifications

Specifications

FRA2
PCRP-FOSL2-1B1
Unconjugated
10 mM PBS with 0.05% BSA
FOS-like antigen 2, fos-related antigen 2, FRA-2, FRA2FLJ23306
Recombinant full-length human FRA2 protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Peptide Array
0.2 mg/ml
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Purified
Mouse
IgG1
100 μg
Cancer, Transcription Factors and Regulators, Tumor Suppressors
Primary
2355
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit