Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fraction V Bovine Albumin, pH 7, Thermo Scientific Chemicals
Heat-Shock fractionated
Supplier: Thermo Scientific J1085722
Description
- Source: Bovine blood
- Form: Lyophilized powder
- Assay (electrophoresis): ≥98.0%
- Protein: ≥96.0%
- pH (2%, H2O): 6.5 - 7.5
- Loss On Drying: ≤5.0%
- Residue on Ignition: ≤2.0%
- CAS: 9048-46-8
Specifications
6.5 to 7.5 | |
Bovine blood | |
Plastic bottle | |
Lyophilized |
100 g | |
4°C | |
Fraction V Bovine Albumin |
Safety and Handling
EINECSNumber : 232-936-2
Recommended Storage : Keep cold
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction