Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Fraction V Bovine Albumin, pH 7

Heat-Shock fractionated

$89.50 - $1075.00

Specifications

Product Type Fraction V Bovine Albumin
Format Lyophilized
Source Bovine blood
Packaging Plastic bottle
pH Range 6.5 - 7.5
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ1085718
View Documents
Thermo Scientific™
J1085718
50g
Each for $89.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ1085722
View Documents
Thermo Scientific™
J1085722
100g
Each for $127.80
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ1085736
View Documents
Thermo Scientific™
J1085736
500g
Each for $705.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ10857A1
View Documents
Thermo Scientific™
J10857A1
1kg
Each for $1,075.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

  • Source: Bovine blood
  • Form: Lyophilized powder
  • Assay (electrophoresis): ≥98.0%
  • Protein: ≥96.0%
  • pH (2%, H2O): 6.5 - 7.5
  • Loss On Drying: ≤5.0%
  • Residue on Ignition: ≤2.0%
  • CAS: 9048-46-8
Specifications

Specifications

Fraction V Bovine Albumin
Bovine blood
6.5 - 7.5
Lyophilized
Plastic bottle
Safety and Handling

Safety and Handling

EINECSNumber : 232-936-2

Recommended Storage : Keep cold

SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit