Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRAS1 (Fraser syndrome 1) Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259101

Catalog No. NBP259101


Add to cart

Description

Description

FRAS1 (Fraser syndrome 1) Polyclonal antibody specifically detects FRAS1 (Fraser syndrome 1) in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

FRAS1 (Fraser syndrome 1)
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
DKFZp686I05113, extracellular matrix protein FRAS1, FLJ14927, FLJ22031, Fraser syndrome 1, KIAA1500DKFZp686P08111
Rabbit
IgG
100 ul
Store at 4C.
Polyclonal
Human
Western Blot, Immunoprecipitation
Western Blot 1:1000, Immunoprecipitation 20 ul/mg lysate
Affinity Purified
FRAS1
Maps to a region between 3025 and 3075
Immunogen affinity purified
RUO
Primary
80144
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only