Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frataxin Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI3C7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270769AF405

Catalog No. N270769A405


Add to cart

Description

Description

Frataxin Monoclonal antibody specifically detects Frataxin in Human samples. It is validated for Western Blot.
Specifications

Specifications

Frataxin
OTI3C7
Western Blot
Affinity Purified, Conjugated
FXN
Full length recombinant protein of human Frataxin (NP_000135) produced in HEK293T cells.
Immunogen affinity purified
Diabetes Research
Primary
2395.0
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit