Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frataxin Mouse anti-Human, Clone: rFXN/2124, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen Frataxin
Clone rFXN/2124
Concentration 0.2 mg/ml
Dilution ELISA 2-4 ug/ml
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123059
View Documents
Novus Biologicals
NBP30779310UG
100 μg
Each for $374.34
Add to cart
 
NB123060
View Documents
Novus Biologicals
NBP30779320UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

Frataxin Monoclonal antibody specifically detects Frataxin in Human samples. It is validated for ELISA
Specifications

Specifications

Frataxin
0.2 mg/ml
ELISA
Purified
RUO
Human
CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199
Recombinant human Frataxin protein fragment (around aa 57-210) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
rFXN/2124
ELISA 2-4 ug/ml
Unconjugated
Mouse
Diabetes Research
10 mM PBS with 0.05% BSA
2395
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit