Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Frataxin Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS9601R

Description
FXN is a mitochondrial protein which belongs to the FRATAXIN family. The protein functions in regulating mitochondrial iron transport and respiration. The expansion of intronic trinucleotide repeat GAA results in Friedreich ataxia.Specifications
Frataxin | |
Polyclonal | |
Unconjugated | |
FXN | |
BOS_8840; CyaY; d-FXN; DKEYP-118A9.1; FA; FARR; frataxin; Frataxin intermediate form; Frataxin mature form; Frataxin(56-210); Frataxin(78-210); Frataxin(81-210); Frataxin, mitochondrial; Frda; Friedreich ataxia; Friedreich ataxia protein; Fxn; i-FXN; m56-FXN; m78-FXN; m81-FXN; RGD1565754; X25; zgc:158218 | |
Rabbit | |
Protein A | |
RUO | |
2395 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q16595 | |
FXN | |
KLH conjugated synthetic peptide derived from human Frataxin. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction