Tocris™ FRAX 486

Potent p21-activated kinase (PAK) inhibitor; brain penetrant and orally bioavailable

Manufacturer: Tocris™ 5190/50

 View more versions of this product

Catalog No. 51-905-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors FRAX 486
    Purity >98
    Molecular Weight 513.39
    Quantity 50mg
    Product Type FRAX 486

    Potent p21-activated kinase (PAK) inhibitor (IC50 values are 14, 33, 39 and 575nM for PAK1, PAK2, PAK3 and PAK4 respectively). Blocks and reverses the DISC1 knockdown-induced reduction in dendritic spine size in cortical neurons. Attenuates dendritic spine elimination and enhances spine generation in DISC1 knockdown mice. Ameliorates autism-like behavioral symptoms in fragile X mental retardation 1 (Fmr1) knockout mice. Brain penetrant and orally bioavailable.

    • Chemical Name: 6-(2,4-Dichlorophenyl)-8-ethyl-2-[[3-fluoro-4-(1-piperazinyl)phenyl]amino]pyrido[2,3-d]pyrimidin-7(8H)-one
    • Formula: C25H23Cl2FN6O
    • CAS Number: 1232030-35-1
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit