Learn More
Carver™ Lab Press Heated Platens: Steel
Manufacturer:  Carver™ 2107 1
Catalog No. 13-872-1
Add to cart
Description
Description
Plastic molding and hot pressing accessory. With control boxes. State preference for thermometer with °C or °F markings.
Specifications
Specifications
Heated Plate
For carver lab press 2089/Model M
Steel
66°C to 260°C
115 V
Control boxes
150°F to 500°F