Learn More
BioVision IncSupplier Diversity Partner Free Glycerol Colorimetric/Fluorometric Assay Kit: Colorimetric & Fluorometric Assay to Measure free Glycerol concentration in various samples within 40 min. Fast, Easy & Sensitive. Detection Range: 50 pmol-10 nmol (1~10000 μM). 100 Assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K630100
Glycerol is widely used in foods, beverages, solvents, pharmaceutical and cosmetic products, etc. There is broad interest in quantification of glycerol for research and development. BioVision’s Glycerol Assay Kit provides a sensitive, easy assay to measure free glycerol concentration in various samples. In the assay, glycerol is enzymatically oxidized to generate a product which reacts with the probe to generate color (λ= 570 nm) and fluorescence (Ex/Em = 535/587 nm). The kit can detect 50 pmol-10 nmol (or 1~10000 µM range) of glycerol in various samples. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9804