Learn More
BioVision IncSupplier Diversity Partner Free Glycerol Colorimetric Assay Kit II: Free Glycerol Colorimetric Assay Kit II: Rapidly Quantitate Free Glycerol in Serum, Tissues, Cells etc. Simple, Sensitive, High-Throughput Adaptable. Detection limit: 20 µM. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K634100
Glycerol is a central component of most lipids. It acts as a backbone for triglycerides and phospholipids, which play an important role in metabolism and cell membrane structure. Due to its low toxicity, glycerol is widely used in the pharmaceutical, food and cosmetic industries. Biovision’s Free Glycerol Assay kit II is suitable for measuring free glycerol levels in samples that contain reducing substances, which may interfere with oxidase-based assays. In this assay, Glycerol in the presence of Glycerol Enzyme Mix is converted to an intermediate, which reduces a colorless Probe to a colored product with strong absorbance at 450 nm. Free Glycerol Assay Kit II is simple, rapid and high-throughput adaptable. This assay kit can detect less than 20 mM of free Glycerol in various biological samples. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9805