Learn More
BioVision IncSupplier Diversity Partner PicoProbe™ Free Glycerol Fluorometric Assay Kit: The most sensitive kit on the market to measure free glycerol. Measures free glycerol in tissues, cells as well as in biological fluids. Detection limit: less than 40 pmol. 100 assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K643100
Glycerol is a central component for synthesis of all lipids; it acts as a backbone for triglycerides and phospholipids, which plays an important role for cell membrane’s structure. Due to its low toxicity, glycerol is widely used in pharmaceutical, food and cosmetic industries. Biovision’s PicoProbeTM Free Glycerol Fluorometric Assay kit is simple, sensitive and easy to use. This assay is suitable for measuring trace amount of glycerol in samples, which contain reducing substances that may interfere with oxidase-based assays. In the assay, glycerol reacts with Enzyme Mix to form an intermediate, which is subsequently oxidized with the production of fluorescence. The fluorescence intensity is directly proportional to the amount of glycerol. This assay kit can detect glycerol amount less than 40 pmol. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9813