Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High-Tech Conversions FREE-SAT™ Presaturated Wipers

High-Tech Conversions FREE-SAT™ Presaturated Wipers are tough, absorbent, and abrasion resistant

Specifications

Autoclavable No
Basis Weight 68 gsm
Color White
Disposable Yes
For Use With (Application) Wiping/Cleaning all aspects of Class 5-8 Cleanrooms
View More Specs

Alert:

Flammable


Products 2
Catalog Number Mfr. No. Saturation Solution Price Quantity & Availability  
Catalog Number Mfr. No. Saturation Solution Price Quantity & Availability  
50-105-6590
View Documents
High-Tech Conversions
FSNT177.9010/DEL
90% IPA / 10% DI water This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
50-105-6587
View Documents
High-Tech Conversions
FSNT177.99/DEL
99% IPA / 1% DI water This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • High level of IPA for fast drying cleanup and extra strength cleaning
  • Pre-saturated with IPA & DI water for added convenience
  • Bagged within a re-sealable container that preserves wipes
  • Poly-Cellulose fabric blend provides a tough, absorbent and abrasion resistant product
Specifications

Specifications

No
White
Wiping/Cleaning all aspects of Class 5-8 Cleanrooms
Poly-Cellulose Blend
Non-sterile
68 gsm
Yes
18 cm
Cleanroom Wiper
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit