Learn More
Chemglass Life SciencesSupplier Diversity Partner Freezer, 13 Cu Ft, Upright, Digital Control, 0 to -40°C DFS Item
Manufacturer:  Chemglass Life SciencesSupplier Diversity Partner CLS3791413
Freezer, 13 Cu Ft, Upright, Digital Control, 0 to -40°C. Our freezer has a digital control with two temperature displays. One display shows the set temperature, while the other shows the actual temperature in the freezer. The freezer has a battery operated alarm system which will emit an audible and visual signal when there is a mechanical or electrical failure. The alarm has an over and under temperature setting, alarm silencing switch and batter test switch. Also supplied with a relay for remote alarm hook-up. The refrigeration system is cascade type with two hermetic compressors supplied with CFC- and HCFC-Free refrigerants. The chamber is 14-ga zinc coated galvanized steel while the exterior is 16-ga steel powder coated with a cool grey finish. ETL Electrical Approved.• Recorder, 7 Day with 6” Chart• Recording Paper, 6; 50 Charts per Pack• Access Openings, 1-2” I.D.• CO² Automatic Back-Up System• Liquid Nitrogen Automatic Back-Up System
Catalog No. 50-143-366