Learn More
Chemglass Life SciencesSupplier Diversity Partner Freezer, 25 Cu Ft, Upright, Digital Control, -40 to -85°C DFS Item
Manufacturer:  Chemglass Life SciencesSupplier Diversity Partner CLS3791825
Freezer, 25 Cu Ft, Upright, Digital Control, -40 to -85°C. Our freezer has a digital control with two temperature displays. One display shows the set temperature, while the other shows the actual temperature in the freezer. The freezer has a battery operated alarm system which will emit an audible and visual signal when there is a mechanical or electrical failure. The alarm has an over and under temperature setting, alarm silencing switch and batter test switch. Also supplied with a relay for remote alarm hook-up. The refrigeration system is cascade type with two hermetic compressors supplied with CFC- and HCFC-Free refrigerants. The chamber is 14-ga zinc coated galvanized steel while the exterior is 16-ga steel powder coated with a cool grey finish. ETL Electrical Approved.
Catalog No. 50-143-510