Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ French Square Bottle, Clear, 1000mL

Click to view available options
Capacity (Metric):
1000 mL

Wheaton French Square bottles are manufactured from clear USP Type III soda-lime glass.

  • Clear USP Type III soda-lime glass
  • Ideal for solvent, chemical or sample storage
  • Square shape maximizes storage space
Closure Included No
Capacity (Metric) 1000 mL
Color Clear
Material Borosilicate Glass (Type 3)
Quantity 24/Case
Type French Square Bottle