Learn More
BlueMed FRESH Shoe Covers
BlueMed FRESH shoe covers are designed to answer the needs for advanced anti-skid and water resistance for a short term use
Manufacturer:  BlueMed SHS1401C
 View more versions of this product
Catalog No. 18-009-112
Add to cart
Description
Description
  • Avanced Anti-Skid Properties
  • Water Resistance
  • Available in Universal and XL Sizes
  • Short Term Use
Specifications
Specifications
Elastic
Universal
Polyethylene, Polypropylene
PPE
17 cm
Yes
Bags
40 gsm
Liquid, skid
Non-sterile
White
Anti-Skid
Yes
6.69 in.
16.93 in.
43 cm
Ultrasonically welded
300 units/case
Shoe cover