Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Freudenberg™ Autoclavable CE Bucket

Blue and red 18 in. autoclavable buckets

Manufacturer:  Freudenberg Household Prod LP 131584

 View more versions of this product

Catalog No. 19-032-1043



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

CE Bucket
6 gal.
6 gal., 22.7L
All popular cleanroom cart systems, 10 or 14 in. Roll-O-Matic™ mops
10 Case
22.7L
Red
Polypropylene
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit