Learn More
Vileda Professional™ EvoControl™ 200 Microfiber Cleanroom Wipes
Designed for fine wiping as it removes contaminants in one swipe. Vileda Professional™ EvoControl™ 200 Microfiber Cleanroom Wipes are widely used in pharmaceutical, biotech and microelectronics industries.
Manufacturer:  Freudenberg Household Prod LP 142855
 View more versions of this product
Catalog No. 19-200-667
  Check Availability
Add to cart
Specifications
Description
  • Clean glass and stainless steel without leaving residues or streaks
  • Microfiber material reach into microscopic surface pores to remove contamination thoroughly
  • Maximize particle pick up
  • Bacteria removal up to 99.999%
  • Cut fibers are melted together for sealed edge which prevents particle release
  • Disposable precision cleaning wipe
  • Laser cut sealed edge wipe for areas where moderate contamination control is required
Description & Specifications
Specifications
ISO 5, LAR, Production and Labs
30.5 x 30.5cm (12 x 12 in.)
Laser cut
150/Pk.; 3000/Cs.
EvoControl 200 Cleanroom Wipes
White
Microfiber