Vileda Professional™ Roll-O-Matic™ Self-Wringing Mops

Suitable for all controlled environments.

Manufacturer: Freudenberg Household Prod LP 118550

 View more versions of this product

Catalog No. 18-999-2407

  Check Availability
Add to Cart

    Specifications

    Color Blue
    For Use With (Application) For use on floors, walls, and ceilings in cleanroom environments. Ideal for the pull-and-lift cleaning technique
    For Use With (Equipment) Roll-O-matic self wringing cleanroom mops
    Includes Wringer built into handle
    Length (English) 58.5 to 76.25 in.
    Width (English) 14 in.
    Autoclavable Yes
    Material Stainless Steel
    Product Type Stainless Steel Mops with Sponge Mop Head, Straight Headl

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit