Freudenberg™ Roll-O-Matic™ Self-Wringing Mops Refills

Suitable for all controlled environments

Manufacturer: Freudenberg Household Prod LP 118808

 View more versions of this product

Catalog No. 18-999-2406

  Check Availability
Add to Cart

    Specifications

    Color Blue
    For Use With (Application) For general use
    For Use With (Equipment) Original and self wringing roll-O-matic roller mops
    Includes Stainless steel channel
    Width (English) 14 in.
    Material Stainless steel
    Product Type Roller Mop Refill

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit