Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5139996

This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: REEHEETQLD IVGIWWTVTN FGEISGTIAI EMDKGTYIH
FRG1 Polyclonal antibody specifically detects FRG1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
FRG1 | |
Polyclonal | |
Unconjugated | |
FRG1 | |
facioscapulohumeral muscular dystrophy region gene-1; Frg1; FRG1A; Frg1l1; FSG1; FSHD region gene 1; FSHD region gene 1 protein; FSHD region gene 1-like 1; hypothetical protein LOC550491; protein FRG1; Unknown (protein for MGC:140313); zgc:112426 | |
Recombinant protein corresponding to Human FRG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
0.1 μg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14331 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2483 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title