Learn More
Friction Rods
Manufacturer: Christensen Educational Materials 9099
Specifications
Friction rod | |
Rods rounded on one end, tapered on other | |
Static electricity, polarity and friction studies |
Lucite | |
0.5 in. | |
12 in. |