Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H-B Instrument™ Frio-Temp™ Digital Verification Thermometers
Calibrated specicifally to verify samples in temperatures in freezers, refrigerators, incubators and ovens.
Supplier: H-B Instrument™ B602102900
Alert:
Plastic bottles as indicated in table cannot be used in ovens. Glass bottle and cap included with products as indicated in the table should not be used in conditions where temperatures may exceed 200°C
Description
- Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
- All models include accredited calibration documentation that meets the requirements of ISO 17025
- Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
- Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish)
- Individually serialized, CE marked
Specifications
Glass Beads | |
4°C Refrigerator | |
0.7 in. | |
Alarm, Timer | |
4.1 in. | |
105 mm | |
2 in. | |
-58°F to +572°F | |
±1°C from -20/130°C; ±2°C from -50/-21°C and 131/200°C; otherwise ±4°C |
30 mL | |
Refrigerator Calibration | |
18 mm | |
3.5 ft. | |
1 m | |
0.1°C/°F; 1 second | |
-50/300°C | |
AAA | |
51 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Includes Prop 65 Cancer and Reproductive Harm warnings
Spot an opportunity for improvement?
Provide Content Correction