Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Frits for Precolumn MicroFilters

Available in either PEEK or Stainless Steel

$90.99 - $106.94

Specifications

Porosity 0.5 µm
Packaging Quantity 10/Pk.
For Use With (Equipment) Precolumn Microfilter Assembly
Diameter (English) Frit 0.031 in.
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
11-100-4573
View Documents
MilliporeSigma™ Supelco™
502790
PEEK
Each for $106.94
Add to cart
 
11-100-4654
View Documents
MilliporeSigma™ Supelco™
502731
Stainless Steel
Each for $90.99
Add to cart
 
Specifications

Specifications

0.5 µm
10/Pk.
Precolumn Microfilter Assembly
0.031 in.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit