Learn More
Frizzled-1 Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0235-9A12, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212316AF532
Catalog No. NP212316A32
Add to cart
Description
Description
Frizzled-1 Monoclonal antibody specifically detects Frizzled-1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Frizzled-1
RM0235-9A12
Western Blot
DKFZp564G072, FLJ95923, frizzled family receptor 1, frizzled homolog 1, frizzled, Drosophila, homolog of, 1, frizzled-1, fz-1, fzE1, hFz1, seven transmembrane spanning receptor, Wnt receptor
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
FZD1
Mouse Frizzled-1
0.1 mL
Primary
8321