Learn More
Frizzled-2 Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0236-7D14, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212317AF594
Catalog No. NP212317A94
Add to cart
Description
Description
Frizzled-2 Monoclonal antibody specifically detects Frizzled-2 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Frizzled-2
RM0236-7D14
Western Blot
frizzled (Drosophila) homolog 2, frizzled homolog 2 (Drosophila), frizzled-2, fz-2, Fz2, FzE2, hFz2
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
FZD2
Mouse Frizzled 2
0.1 mL
Primary
2535