Learn More
Frizzled-6 Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0237-6H38, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212318AF750
Catalog No. NP212318A70
Add to cart
Description
Description
Frizzled-6 Monoclonal antibody specifically detects Frizzled-6 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Frizzled-6
RM0237-6H38
Western Blot
frizzled (Drosophila) homolog 6, frizzled homolog 6 (Drosophila), frizzled-6, fz-6, FZ6, HFZ6, seven transmembrane helix receptor
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
FZD6
Mouse Frizzled-6
0.1 mL
Primary
8323