Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Frizzled-6 Rat anti-Mouse, Clone: RM0237-6H38, Novus Biologicals™
Rat Monoclonal Antibody
Specifications
Antigen | Frizzled-6 |
---|---|
Clone | RM0237-6H38 |
Concentration | LYOPH |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP212318
|
Novus Biologicals
NBP212318 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB019168
|
Novus Biologicals
NBP2123180.025MG |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Frizzled-6 Monoclonal antibody specifically detects Frizzled-6 in Mouse samples. It is validated for Western Blot.Specifications
Frizzled-6 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway | |
Q61089 | |
8323 | |
Mouse Frizzled-6 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RM0237-6H38 | |
Western Blot 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
frizzled (Drosophila) homolog 6, frizzled homolog 6 (Drosophila), frizzled-6, fz-6, FZ6, HFZ6, seven transmembrane helix receptor | |
FZD6 | |
IgG2 | |
Protein G purified |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title