Learn More
Frizzled-7 Rat anti-Mouse, Alexa Fluor™ 350, Clone: RM0238-15B7, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212319AF350
Catalog No. NP212319A30
Add to cart
Description
Description
Frizzled-7 Monoclonal antibody specifically detects Frizzled-7 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Frizzled-7
RM0238-15B7
Western Blot
frizzled (Drosophila) homolog 7, frizzled homolog 7 (Drosophila), Frizzled, drosophila, homolog of, 7, Fz-7, FzE3frizzled-7, hFz7
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
FZD7
Mouse Frizzled-7
0.1 mL
Primary
8324