Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frizzled-8 Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0239-11G21, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212320AF532

Catalog No. NP212320A32


Add to cart

Description

Description

Frizzled-8 Monoclonal antibody specifically detects Frizzled-8 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Frizzled-8
RM0239-11G21
Western Blot
Conjugated, Purified
FZD8
Mouse Frizzled-8
Protein G purified
GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway
Primary
8325.0
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
frizzled (Drosophila) homolog 8, frizzled 8, frizzled family receptor 8, frizzled homolog 8 (Drosophila), frizzled-8, FZ-8, hFz8
Rat
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit