Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Frizzled-8 Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0239-11G21, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212320AF750
Catalog No. NP212320A70
Description
Description
Frizzled-8 Monoclonal antibody specifically detects Frizzled-8 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Frizzled-8
RM0239-11G21
Western Blot
frizzled (Drosophila) homolog 8, frizzled 8, frizzled family receptor 8, frizzled homolog 8 (Drosophila), frizzled-8, FZ-8, hFz8
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
FZD8
Mouse Frizzled-8
0.1 mL
Primary
8325