Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frk Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI6D2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270775AF405

Catalog No. N270775A405


Add to cart

Description

Description

Frk Monoclonal antibody specifically detects Frk in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Frk
OTI6D2
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
FRK
Full length recombinant protein of human Frk (NP_002022) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication
Primary
2444.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, fyn-related kinaseProtein-tyrosine kinase 5, GTKEC 2.7.10.2, Nuclear tyrosine protein kinase RAK, PTK5 protein tyrosine kinase 5, RAKPTK5, tyrosine-protein kinase FRK
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit