Learn More
FRK Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549805
Catalog No. PA549805
Add to cart
Description
Description
The antibody detects endogenous levels of total FRK protein.
FRK (Fyn-related kinase), also known as Rak. It is an epithelial tissue-specific kinase. The human FRK gene maps to chromosome 6q21-q22.3 and encodes a 505 amino acid protein. The protein belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth.
Specifications
Specifications
FRK
Polyclonal
Unconjugated
FRK
Liquid
FRK
Synthesized peptide derived from internal of human FRK.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P42685, Q922K9, Q62662
EC 2.7.1.112, EC 2.7.10.2, Nuclear tyrosine protein kinase RAK, Tyrosine-protein kinase FRK, fyn-related kinase
Rabbit
IgG
100 μL
-20°C
Primary
14302, 2444, 79209