Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRK Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549385

Catalog No. PA549385

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth.
Specifications

Specifications

Frk
Polyclonal
PBS with 0.09% sodium azide
P42685, Q922K9, Q62662
Tyrosine-protein kinase FRK, Beta-cell Src-homology tyrosine kinase, BSK, FYN-related kinase, Intestine tyrosine kinase, Frk, Bsk, Iyk
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14302, 2444, 79209
Western Blot
Unconjugated
FRK
Liquid
FRK
KLH conjugated synthetic peptide between 482-516 amino acids from the C-terminal region of human Mouse Frk.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit