Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRMD3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549034

Catalog No. PA549034

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Putative tumor suppressor gene that may be implicated in the origin and progression of lung cancer.
Specifications

Specifications

FRMD3
Polyclonal
PBS with 0.09% sodium azide
A2A2Y4
FERM domain-containing protein 3, Band 41-like protein 4O, Ovary type protein 41, 41O, FRMD3, EPB41L4O
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
257019
Western Blot
Unconjugated
FRMD3
Liquid
FRMD3
KLH conjugated synthetic peptide between 250-279 amino acids from the Central region of human FRMD3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit