Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FRMD6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569884
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
FRMD6 | |
Polyclonal | |
Unconjugated | |
Frmd6 | |
2610019M19Rik; 4.1 ezrin radixin moesin (FERM)-containing protein; 4930488L10Rik; AW212977; c14_5320; C14orf31; EX1; expanded homolog; FERM domain containing 6; FERM domain containing 6; FERM domain-containing protein 6; FERM domain-containing protein 163SCII; FERM domain-containing protein 6; FERM-domain-containing protein 163SCII; Frmd6; MGC17921; Willin | |
Rabbit | |
Affinity chromatography | |
RUO | |
257646 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8VII0 | |
Frmd6 | |
synthetic peptide directed towards the C-terminal region of rat FRMD6. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction