Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRMD7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548565

Catalog No. PA548565

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Plays a role in neurite development, may be through the activation of the GTPase RAC1. Plays a role in the control of eye movement and gaze stability.
Specifications

Specifications

FRMD7
Polyclonal
PBS with 0.09% sodium azide
Q6ZUT3
FERM domain-containing protein 7, FRMD7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
90167
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FRMD7
Liquid
FRMD7
KLH conjugated synthetic peptide between 115-144 amino acids from the N-terminal region of human FRMD7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit