Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DeltaBio Frogs, Bio-Fresh™
SCI_ED

Bio-Fresh™ frog specimens are fixed in formaldehyde, repeatedly rinsed and packaged in Bio-Fresh solution. These budget-friendly specimens meet or exceed OSHA lab standards for formaldehyde, and are available with colored latex injections.

 View more versions of this product

Catalog No. S160150

Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.


Description

Description

INJECTION KEY

  • Plain: Circulatory system not injected with colors
  • Single: Arteries injected with red latex
  • Double: Arteries red, veins blue
  • Triple: Arteries red, veins blue and hepatic portal veins yellow

INJECTION KEY
  • Plain: Circulatory system not injected with colors
  • Single: Arteries injected with red latex
  • Double: Arteries red, veins blue
  • Triple: Arteries red, veins blue and hepatic portal veins yellow
Specifications

Specifications

Grassfrog
Plain
7.62 to 8.89 cm
Pail
Bio-Fresh™
3 to 3.5 in.
50/Pail
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit