Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frogs, Bio-Fresh™ Products for Science Education

Bio-Fresh™ frog specimens are fixed in formaldehyde, repeatedly rinsed and packaged in Bio-Fresh solution. These budget-friendly specimens meet or exceed OSHA lab standards for formaldehyde, and are available with colored latex injections.

 View more versions of this product

Catalog No. S160110


Add to cart

Description

Description

INJECTION KEY
  • Plain: Circulatory system not injected with colors
  • Single: Arteries injected with red latex
  • Double: Arteries red, veins blue
  • Triple: Arteries red, veins blue and hepatic portal veins yellow
Specifications

Specifications

Education
3 to 3.5 in.
Pail
Grassfrog
Plain
7.62 to 8.89cm
Bio-Fresh™
10/Pail
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit