Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fructosamine-3-kinase-related Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | Fructosamine-3-kinase-related |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP156367
|
Novus Biologicals
NBP156367 |
100 μL |
Each of 1 for $436.00
|
|
Description
Fructosamine-3-kinase-related Polyclonal specifically detects Fructosamine-3-kinase-related in Human samples. It is validated for Western Blot.Specifications
Fructosamine-3-kinase-related | |
Polyclonal | |
Rabbit | |
Protein Kinase | |
Q9HA64 | |
79672 | |
Synthetic peptides corresponding to FN3KRP(fructosamine-3-kinase-related protein) The peptide sequence was selected from the N terminal of FN3KRP. Peptide sequence MDPGDPAGDPAAGERHRMGRDPLLLLQALQTLWSTRERKQLREEAWRGFA. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
EC 2.7.1.-, FLJ12171, FN3KL, FN3K-related protein, FN3K-RP, fructosamine 3 kinase related protein, Fructosamine-3-kinase-related protein, ketosamine-3-kinase | |
FN3KRP | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title