Learn More
BioVision IncSupplier Diversity Partner Fructose Colorimetric/Fluorometric Assay Kit: Fructose Assay Kit: High-Throughput Suitable Colorimetric & Fluorometric Assay to Measure Fructose in a variety of Biological Samples. Rapid, Simple & Sensitive. 100 Assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K619100
Fructose is a monosaccharide found in many foods and is one of the three most important blood sugars along with glucose and galactose. Fructose is the sweetest naturally occurring sugar, estimated to be twice as sweet as sucrose. In BioVision’s Fructose Assay Kit, free fructose is enzymatically converted to β-glucose, which is then specifically converted to a product that reacts with OxiRed Probe to generate color (λ=570nm) and fluorescence (Ex/Em=535/587nm). The kit provides a rapid, simple, sensitive, and reliable method suitable for high throughput assayof D-fructose. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9800