Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSH beta Mouse anti-Human, PE/Atto594, Clone: OTI2A6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270781PEATT594

Catalog No. N270781P594


Add to cart

Description

Description

FSH beta Monoclonal antibody specifically detects FSH beta in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

FSH beta
OTI2A6
Flow Cytometry
Affinity Purified, Conjugated
FSHB
Full length recombinant protein of human FSH beta (NP_001018090) produced in HEK293T cells.
Immunogen affinity purified
Cancer
Primary
2488.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
follicle stimulating hormone, beta polypeptide, Follicle-stimulating hormone beta subunit, Follitropin beta chain, follitropin subunit beta, follitropin, beta chain, FSH-B, FSH-beta
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit