Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FSHR Mouse anti-Human, Alexa Fluor 647, Clone: 626717, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB65591R100UG
This item is not returnable.
View return policy
Description
FSHR Monoclonal specifically detects FSHR in Human samples. It is validated for Flow Cytometry.Specifications
FSHR | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
follicle stimulating hormone receptor, follicle-stimulating hormone receptor, Follitropin receptor, FSH R, FSH receptor, FSH-R, LGR1, LGR1FSHRO, MGC141667, MGC141668, ODG1 | |
NS0 mouse myeloma cell line transfected with human FSH R Accession # M65085 | |
100 μg | |
2492 | |
Human | |
Supernatant |
Flow Cytometry | |
626717 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
M65085 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human FSH R in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title