Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSHR Mouse anti-Human, Alexa Fluor™ 350, Clone: FSHR/1400, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254329AF350

Catalog No. NP24329A30


Add to cart

Description

Description

FSHR Monoclonal antibody specifically detects FSHR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

FSHR
FSHR/1400
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Conjugated, Purified
FSHR
Recombinant human full-length FSHR protein
Protein A or G purified
GPCR
Primary
2492.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
follicle stimulating hormone receptor, follicle-stimulating hormone receptor, Follitropin receptor, FSH receptor, FSH-R, LGR1FSHRO, MGC141667, MGC141668, ODG1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit