Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FSHR Antibody (6E8.2F5), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP236489AF750
Description
FSHR Monoclonal specifically detects FSHR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FSHR | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
FSHR | |
A synthetic peptide made to the human FSH R protein sequence (between residues 1-200). [Swiss-Prot P23945] | |
0.1 mL | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
6E8.2F5 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
follicle stimulating hormone receptor, follicle-stimulating hormone receptor, Follitropin receptor, FSH receptor, FSH-R, LGR1FSHRO, MGC141667, MGC141668, ODG1 | |
Mouse | |
Protein G purified | |
GPCR | |
2492.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction